Learn More
Biomatik Corporation Dipalmitin (PP) ELISA Kit
Supplier:  Biomatik Corporation EKU10795
Encompass_Preferred
Dipalmitin (PP) ELISA Kit for detecting PP in biological agents. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 1,3-Dipalmitin; 1,3-Di-(Hexadecanoyl)-Glycerol; 1,3-Dipalmitoylglycerol; 1,3-Dipalmitoyl-Glycerol, Glyceryl 1,3-Dipalmitate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4896