Learn More
Echelon Biosciences Research Labs DIPALMITOYL-SN-GLYCEROL 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L0016250MG
Encompass
1,2-Diacylglycerols (DAGs) are formed in cells through the hydrolysis of phosphatidylinositol 4,5-bisphosphate by Phospholipase C. DAGs activate Protein Kinase C (PKC), which is a key regulator of signal transduction, cellular regulation, and tumor promotion. This item is non returnable
Catalog No. 50-720-464