Learn More
Medchemexpress LLC Dipalmitoylphosphatidylcholine-d6 (racemic) | 82765-17-1 | 98.0% | 740.08 g/mol | C40H74D6NO8P | 5 MG
SDP
Catalog No. 5000263410
Encompass_Preferred
Deuterated dipalmitoylphosphatidylcholine (racemic) (DPPC-d6) is an isotopically labeled phosphoglyceride supplied as a solid for use as an analytical standard and for preparing lipid monolayers, bilayers, and liposomes. It is used in biophysical experiments and as a tracer in mass-spectrometry and formulation studies. Certificate of analysis reports HPLC purity of 98.0% and the material is typically stored sealed at -20 °C.
  • Isotopically labeled (D6) for tracer and mass-spectrometry applications.
  • Suitable for preparing lipid monolayers, bilayers, and liposomes.
  • High HPLC purity for reliable analytical results.
  • Supplied as a stable powder for sealed cold storage.
  • Useful as an internal standard in quantitative analyses.
Catalog No. 50-002-63410 Supplier Medchemexpress LLC Supplier No. HY109506S25MG
Deuterated dipalmitoylphosphatidylcholine (racemic) (DPPC-d6) is an isotopically labeled phosphoglyceride supplied as a solid for use as an analytical standard and for preparing lipid monolayers, bilayers, and liposomes. It is used in biophysical experiments and as a tracer in mass-spectrometry and formulation studies. Certificate of analysis reports HPLC purity of 98.0% and the material is typically stored sealed at -20 °C.
  • Isotopically labeled (D6) for tracer and mass-spectrometry applications.
  • Suitable for preparing lipid monolayers, bilayers, and liposomes.
  • High HPLC purity for reliable analytical results.
  • Supplied as a stable powder for sealed cold storage.
  • Useful as an internal standard in quantitative analyses.