Learn More
Medchemexpress LLC Dipalmitoylphosphatidylcholine-d62 | 25582-63-2 | MFCD00151588 | 99.0% | 796.42 g·mol⁻¹ | C40H18D62NO8P | 5 MG
SDP
Catalog No. 5000263635
Encompass_Preferred
DPPC-d62 is the deuterium-labeled form of dipalmitoylphosphatidylcholine (DPPC) used to prepare lipid monolayers, bilayers, and liposomes. The deuterium label enables stable-isotope tracing and spectroscopic differentiation from non-labeled lipids, making the compound useful in biophysical, analytical, and formulation studies. Supplied as a powder, it is intended for in vitro and appropriate in vivo preparations.
Catalog No. 50-002-63635 Supplier Medchemexpress LLC Supplier No. HY109506S5MG
DPPC-d62 is the deuterium-labeled form of dipalmitoylphosphatidylcholine (DPPC) used to prepare lipid monolayers, bilayers, and liposomes. The deuterium label enables stable-isotope tracing and spectroscopic differentiation from non-labeled lipids, making the compound useful in biophysical, analytical, and formulation studies. Supplied as a powder, it is intended for in vitro and appropriate in vivo preparations.