Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentaerythritol Hexakis(3-mercaptopropionate) 93.0+%, TCI America™
SDP

Catalog No. D5212500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5212500G 500 g
D521225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5212500G Supplier TCI America Supplier No. D5212500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25359-71-1
Molecular Formula C28H46O13S6
Molecular Weight (g/mol) 783.023
MDL Number MFCD00128217
InChI Key YAAUVJUJVBJRSQ-UHFFFAOYSA-N
Synonym DiPETMP
PubChem CID 117451
IUPAC Name [3-(3-sulfanylpropanoyloxy)-2-[[3-(3-sulfanylpropanoyloxy)-2,2-bis(3-sulfanylpropanoyloxymethyl)propoxy]methyl]-2-(3-sulfanylpropanoyloxymethyl)propyl] 3-sulfanylpropanoate
SMILES C(CS)C(=O)OCC(COCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

Specifications

Color Yellow
Quantity 500 g
Formula Weight 783.02
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material Dipentaerythritol Hexakis(3-mercaptopropionate)
EINECSNumber (2)-3335
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.