Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dipentaerythritol penta-/hexa-acrylate

Catalog No. 407283500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-728-3500ML 500 mL
40-728-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-728-3500ML Supplier Sigma Aldrich Supplier No. 407283500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dipentaerythritol penta-/hexa-acrylate (DPHPA) is a multifunctional acrylate.DPHPA may be used to prepare nanoporous/non porous polymeric thin films for drug delivery systems. One of the constituents of prepolymer syrup to fabricate holographic polymer-dispersed liquid crystals(H-PDLCs) gratings is DPHPA . It may be used to form a photopolymer blend for various applications such as microelectronics and micro-optical elements in displays.

Specifications

CAS 60506-81-2
Density 1.155 g/mL (at 25°C)
Molecular Formula C25H32O12
Refractive Index n20/D 1.49 (literature)
Linear Formula O[CH2C(CH2OR)3]2, R = COCH=CH2 or H
MDL Number MFCD00192104
Quantity 500 mL
Synonym Dipentaerythritol monohydroxypentaacrylate; Dipentaerythritol pentaacrylate
Molecular Weight (g/mol) 524.51
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.