Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich dipentamethylenethiuram tetrasulfide

Catalog No. CDS0105841G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS0105841G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS0105841G Supplier Sigma Aldrich Supplier No. CDS0105841G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,nα-diethylethylenediamine has been used as an internal standard in selective liquid chromatographic method developed for the assay of ethambutol in serum samples
  • It has also been used for the solvation of lithium hexamethyldisilazide

Specifications

Molecular Formula C12H20N2S6
Linear Formula C12H20N2S6
MDL Number MFCD00047474
Quantity 25 g
Molecular Weight (g/mol) 384.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.