Learn More
Medchemexpress LLC Dipentyl phthalate | 131-18-0 | 99.4% | 306.40 | 10MM/1ML
SDP
Catalog No. 5000454542
Encompass_Preferred
Dipentyl phthalate is an endocrine-disrupting phthalate plasticizer that increases AMPK phosphorylation and decreases AKT1 phosphorylation and SIRT1 levels. It also reduces adrenocorticotropic hormone levels and is considered a testicular toxicant.
  • Increases AMPK phosphorylation
  • Decreases AKT1 phosphorylation
  • Decreases SIRT1 levels
  • Reduces adrenocorticotropic hormone levels
  • Functions as an endocrine-disrupting phthalate plasticizer
  • Considered a testicular toxicant
Catalog No. 50-004-54542 Supplier Medchemexpress LLC Supplier No. HYW01381610MM/1ML
Dipentyl phthalate is an endocrine-disrupting phthalate plasticizer that increases AMPK phosphorylation and decreases AKT1 phosphorylation and SIRT1 levels. It also reduces adrenocorticotropic hormone levels and is considered a testicular toxicant.
  • Increases AMPK phosphorylation
  • Decreases AKT1 phosphorylation
  • Decreases SIRT1 levels
  • Reduces adrenocorticotropic hormone levels
  • Functions as an endocrine-disrupting phthalate plasticizer
  • Considered a testicular toxicant