Learn More
Medchemexpress LLC Dipentyl phthalate-3,4,5,6-d4 | 358730-89-9 | 99.3% | 310.42 | 1 MG
SDP
Catalog No. 5000454488
Encompass_Preferred
Dipentyl phthalate-3,4,5,6-d4 is the deuterium labeled Dipentyl phthalate-3,4,5,6. It is for research use only. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54488 Supplier Medchemexpress LLC Supplier No. HYW013816S1MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dipentyl phthalate-3,4,5,6-d4 is the deuterium labeled Dipentyl phthalate-3,4,5,6. It is for research use only. Deuteration, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.