Learn More
Medchemexpress LLC Dipentyl phthalate-3,4,5,6-d4 | 358730-89-9 | 99.3% | 310.42 | 5 MG
SDP
Catalog No. 5000454650
Encompass_Preferred
Dipentyl phthalate-3,4,5,6-d4 is the deuterium labeled Dipentyl phthalate-3,4,5,6. It is intended for research use only and is not for sale to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54650 Supplier Medchemexpress LLC Supplier No. HYW013816S5MG
Dipentyl phthalate-3,4,5,6-d4 is the deuterium labeled Dipentyl phthalate-3,4,5,6. It is intended for research use only and is not for sale to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS