Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111019004 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9004 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9004 Supplier MilliporeSigma Supelco Supplier No. 3419410MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 358730-89-9
Flash Point 118°C - Closed Cup
Physical Form Neat
Linear Formula C18D4H22O4
MDL Number MFCD01861607
Synonym Di-n-pentyl phthalate-d4
Type Analytical Standard
Quantity 10 mg
Formula Weight 310.42
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.