Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentylamine 99%, Thermo Scientific™

Catalog No. AC219360100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC219360100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219360100 Supplier Thermo Scientific Chemicals Supplier No. 219360100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-92-2
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.301
InChI Key JACMPVXHEARCBO-UHFFFAOYSA-N
Synonym dipentylamine, diamylamine, di-n-amylamine, 1-pentanamine, n-pentyl, di-n-pentylamine, diamyl amine, pentylamine, pentyl, amine, dipentyl, unii-d67i8pn8uu, ccris 6225
PubChem CID 16316
IUPAC Name N-pentylpentan-1-amine
SMILES CCCCCNCCCCC

Specifications

Color Colorless to Yellow
Density 0.77
Infrared Spectrum Authentic
Refractive Index 1.4262 to 1.4282 (20°C, 589nm)
Linear Formula [CH3(CH2)4]2NH
Quantity 10 mL
Beilstein 04, I, 378
Formula Weight 157.3
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Dipentylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.