Learn More
Cayman Chemical Dipeptide diaminobutyroyl benzylamide (hydrochloride), Purity: ≥98%, 250 mg
SDP
Catalog No. 502941558
Encompass_Preferred
A biomimetic peptide and muscarinic acetylcholine receptor antagonist; blocks sodium uptake and induces muscle relaxation
Catalog No. 50-294-1558 Supplier Cayman Chemical Supplier No. 27566250 MG
A biomimetic peptide and muscarinic acetylcholine receptor antagonist; blocks sodium uptake and induces muscle relaxation