Learn More
Cayman Chemical Dipeptide diaminobutyroyl benzylamide (hydrochloride), Purity: ≥98%, 100 mg
SDP
Catalog No. 502941302
Encompass_Preferred
A biomimetic peptide and muscarinic acetylcholine receptor antagonist; blocks sodium uptake and induces muscle relaxation
Catalog No. 50-294-1302 Supplier Cayman Chemical Supplier No. 27566100 MG
A biomimetic peptide and muscarinic acetylcholine receptor antagonist; blocks sodium uptake and induces muscle relaxation