Learn More
Biomatik Corporation Dipeptidase 1, Renal (Dpep1) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30780
Encompass_Preferred
Dipeptidase 1, Renal (Dpep1) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Dipeptidase 1, Renal (DPEP1). The Dipeptidase 1, Renal (Dpep1) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp17~Leu411. Applications: WB. Target Synonyms: Dipeptidase 1, Renal (DPEP1). Accession Number: P16444. Gene ID: 1800. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6466