Learn More
Cusabio Technology LLC dipeptidyl-peptidase 4
Catalog No. 5000048472
Encompass_Preferred
This kit detects DPP4 in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/ml to 20 ng/ml with a sensitivity of 0 078 ng/ml ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 l and can be completed within 1-5 hours Optical density is measured at 450 nm for accurate quantification Designed for researchers requiring reliable and reproducible results this kit is optimized for both diagnostic and experimental applications involving DPP4 analysis
Catalog No. 50-000-48472 Supplier Cusabio Technology LLC Supplier No. CSBE08520M
This kit detects DPP4 in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/ml to 20 ng/ml with a sensitivity of 0 078 ng/ml ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 l and can be completed within 1-5 hours Optical density is measured at 450 nm for accurate quantification Designed for researchers requiring reliable and reproducible results this kit is optimized for both diagnostic and experimental applications involving DPP4 analysis