Learn More
Medchemexpress LLC Diperdipine | 108852-42-2 | 457.52 | 1 ML
SDP
Catalog No. 5000448440
Encompass_Preferred
YS-201 is a dihydropyridine-type calcium channel antagonist with potential for treating angina pectoris and hypertension. In vivo, YS-201 (Diperdipine) significantly reduces systemic vascular resistance and improves stroke index and left ventricular ejection fraction.
  • Dihydropyridine-type calcium channel antagonist
  • Potential for treating angina pectoris and hypertension
  • Significantly reduces systemic vascular resistance in vivo
  • Improves stroke index and left ventricular ejection fraction in vivo
  • Absolute bioavailability is 18.7% after intravenous and oral doses
  • Bioavailable fraction increases up to 44.3% after intraportal administration
Catalog No. 50-004-48440 Supplier Medchemexpress LLC Supplier No. HYU0013710MM/1ML
YS-201 is a dihydropyridine-type calcium channel antagonist with potential for treating angina pectoris and hypertension. In vivo, YS-201 (Diperdipine) significantly reduces systemic vascular resistance and improves stroke index and left ventricular ejection fraction.
  • Dihydropyridine-type calcium channel antagonist
  • Potential for treating angina pectoris and hypertension
  • Significantly reduces systemic vascular resistance in vivo
  • Improves stroke index and left ventricular ejection fraction in vivo
  • Absolute bioavailability is 18.7% after intravenous and oral doses
  • Bioavailable fraction increases up to 44.3% after intraportal administration