Learn More
Medchemexpress LLC Diperdipine | 108852-42-2 | 457.52 | 10 MG
SDP
Catalog No. 5000448885
Encompass_Preferred
YS-201 (Diperdipine) is a dihydropyridine-type calcium channel antagonist that targets calcium channels. It is involved in membrane transporter/ion channel and neuronal signaling pathways and has potential for angina pectoris and hypertension treatment.
  • Functions by blocking calcium channels.
  • Markedly reduces systemic vascular resistance.
  • Improves stroke index and left ventricular ejection fraction.
  • Leads to an augmented left ventricular end-diastolic volume index, indicating an increase in preload.
  • Has an absolute bioavailability of 18.7% after intravenous and oral doses.
  • Applicable in the treatment of angina pectoris and hypertension.
Catalog No. 50-004-48885 Supplier Medchemexpress LLC Supplier No. HYU0013710MG
YS-201 (Diperdipine) is a dihydropyridine-type calcium channel antagonist that targets calcium channels. It is involved in membrane transporter/ion channel and neuronal signaling pathways and has potential for angina pectoris and hypertension treatment.
  • Functions by blocking calcium channels.
  • Markedly reduces systemic vascular resistance.
  • Improves stroke index and left ventricular ejection fraction.
  • Leads to an augmented left ventricular end-diastolic volume index, indicating an increase in preload.
  • Has an absolute bioavailability of 18.7% after intravenous and oral doses.
  • Applicable in the treatment of angina pectoris and hypertension.