Learn More
Selleck Chemical LLC Diphemanil Methylsulfate S4034-50mg
SDP
Catalog No. 503108400
Encompass_Preferred
Diphemanil Methylsulfate is a quaternary ammonium anticholinergic it binds muscarinic acetycholine receptors (mAchR)
Catalog No. 50-310-8400 Supplier Selleck Chemical LLC Supplier No. S403450MG
Diphemanil Methylsulfate is a quaternary ammonium anticholinergic it binds muscarinic acetycholine receptors (mAchR)