Learn More
Cayman Chemical Diphencyprone, Purity: ≥95%, CAS: 886-38-4, 500 mg
SDP
Catalog No. 502941157
Encompass_Preferred
A contact sensitizing agent; pretreatment reduces symptom severity in a mouse model of MBP1-10-induced EAE; enhances IL-10 production and antigen-specific IgG2a antibody responses and reduces anaphylaxis and asthma induced by ovalbumin in mice at 1% v/v; topical administration increases intrafollicular CD4+ and CD8+ T cells, reduces upper dermal inflammation, and stimulates hair growth in the C3H/HeJ mouse model of alopecia areta
Catalog No. 50-294-1157 Supplier Cayman Chemical Supplier No. 27982500 MG
A contact sensitizing agent; pretreatment reduces symptom severity in a mouse model of MBP1-10-induced EAE; enhances IL-10 production and antigen-specific IgG2a antibody responses and reduces anaphylaxis and asthma induced by ovalbumin in mice at 1% v/v; topical administration increases intrafollicular CD4+ and CD8+ T cells, reduces upper dermal inflammation, and stimulates hair growth in the C3H/HeJ mouse model of alopecia areta