Learn More
Medchemexpress LLC Diphenhydramine-d5 hydrochloride | 1219795-16-0 | 99.8% | 5 MG
SDP
Catalog No. 5000405464
Encompass_Preferred
Diphenhydramine-d5 hydrochloride is a deuterium-labeled Diphenhydramine hydrochloride. It is a first-generation histamine H1-receptor antagonist with anti-cholinergic effects and can cross the ovine blood-brain barrier (BBB). This compound is intended for research use only.
  • Deuterium labeled Diphenhydramine hydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05464 Supplier Medchemexpress LLC Supplier No. HYB0303AS15MG
Diphenhydramine-d5 hydrochloride is a deuterium-labeled Diphenhydramine hydrochloride. It is a first-generation histamine H1-receptor antagonist with anti-cholinergic effects and can cross the ovine blood-brain barrier (BBB). This compound is intended for research use only.
  • Deuterium labeled Diphenhydramine hydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS