Learn More
Medchemexpress LLC Diphenhydramine-d5 hydrochloride | 1219795-16-0 | MFCD12022195 | 99.7% | 296.85 | C17H17D5ClNO | 10 MG
SDP
Catalog No. 5000299006
Encompass_Preferred
Diphenhydramine-d5 hydrochloride is a deuterium-labeled hydrochloride salt of diphenhydramine supplied as a research-grade reference standard for analytical and pharmacokinetic studies. It is intended for use as an internal standard in mass spectrometry and related assays; not for human consumption.
  • deuterium-labeled (d5) analogue for tracer and internal standard use.
  • hydrochloride salt form for stability and solubility in analytical workflows.
  • suitable for LC-MS/MS and quantitative pharmacokinetic studies.
  • low-quantity packaging for analytical calibration and method development.
  • molecular weight 296.85 and CAS 1219795-16-0 for unambiguous identification.
Catalog No. 50-002-99006 Supplier Medchemexpress LLC Supplier No. HYB0303AS110MG
Diphenhydramine-d5 hydrochloride is a deuterium-labeled hydrochloride salt of diphenhydramine supplied as a research-grade reference standard for analytical and pharmacokinetic studies. It is intended for use as an internal standard in mass spectrometry and related assays; not for human consumption.
  • deuterium-labeled (d5) analogue for tracer and internal standard use.
  • hydrochloride salt form for stability and solubility in analytical workflows.
  • suitable for LC-MS/MS and quantitative pharmacokinetic studies.
  • low-quantity packaging for analytical calibration and method development.
  • molecular weight 296.85 and CAS 1219795-16-0 for unambiguous identification.