Learn More
Apexbio Technology LLC Diphenhydramine HCl 147-24-0 500mg
SDP
Catalog No. 5000089969
Encompass_Preferred
Diphenhydramine HCl is a small-molecule antagonist targeting histamine H1 receptors It is designed to competitively inhibit histamine binding to H1 receptors in peripheral tissues and the central nervous system thereby suppressing histamine-mediated hypersensitivity responses Diphenhydramine HCl exerts its biological activity primarily through antagonism of histamine H1 receptors and also exhibits anticholinergic activity within the central nervous system Based on these pharmacological properties Diphenhydramine HCl holds research potential in the study of allergic reactions sleep regulation sedation pathways motor disorders cognitive function memory processes and investigations of cholinergic and histaminergic neurotransmission
Catalog No. 50-000-89969 Supplier Apexbio Technology LLC Supplier No. B1932500
Diphenhydramine HCl is a small-molecule antagonist targeting histamine H1 receptors It is designed to competitively inhibit histamine binding to H1 receptors in peripheral tissues and the central nervous system thereby suppressing histamine-mediated hypersensitivity responses Diphenhydramine HCl exerts its biological activity primarily through antagonism of histamine H1 receptors and also exhibits anticholinergic activity within the central nervous system Based on these pharmacological properties Diphenhydramine HCl holds research potential in the study of allergic reactions sleep regulation sedation pathways motor disorders cognitive function memory processes and investigations of cholinergic and histaminergic neurotransmission