Learn More
Medchemexpress LLC HY-B0303A 250mg , Diphenhydramine (hydrochloride) CAS:147-24-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0303A250MG
Encompass_Preferred
HY-B0303A 250mg Diphenhydramine (hydrochloride) CAS: 147-24-0 Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effect. Diphenhydramine hydrochloride can across the ovine blood-brain barrier (BBB). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1127