Learn More
Apexbio Technology LLC Diphenhydramine HCl 147-24-0 250mg
SDP
Catalog No. 502240668
Encompass_Preferred
Diphenhydramine HCl is a small-molecule antagonist targeting histamine H1 receptors It is designed to competitively inhibit histamine binding to H1 receptors in peripheral tissues and the central nervous system thereby suppressing histamine-mediated hypersensitivity responses Diphenhydramine HCl exerts its biological activity primarily through antagonism of histamine H1 receptors and also exhibits anticholinergic activity within the central nervous system Based on these pharmacological properties Diphenhydramine HCl holds research potential in the study of allergic reactions sleep regulation sedation pathways motor disorders cognitive function memory processes and investigations of cholinergic and histaminergic neurotransmission
This item is not returnable. View return policy
Catalog No. 50-224-0668 Supplier Apexbio Technology LLC Supplier No. B1932250
Diphenhydramine HCl is a small-molecule antagonist targeting histamine H1 receptors It is designed to competitively inhibit histamine binding to H1 receptors in peripheral tissues and the central nervous system thereby suppressing histamine-mediated hypersensitivity responses Diphenhydramine HCl exerts its biological activity primarily through antagonism of histamine H1 receptors and also exhibits anticholinergic activity within the central nervous system Based on these pharmacological properties Diphenhydramine HCl holds research potential in the study of allergic reactions sleep regulation sedation pathways motor disorders cognitive function memory processes and investigations of cholinergic and histaminergic neurotransmission