Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine Hydrochloride, 99.32%, MP Biomedicals™

Catalog No. ICN19004480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19004480 100 g
ICN19004410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19004480 Supplier MP Biomedicals, Inc Supplier No. 0219004480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number MFCD00012479
InChI Key PCHPORCSPXIHLZ-UHFFFAOYSA-N
Synonym diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
PubChem CID 8980
ChEBI CHEBI:4637
IUPAC Name hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
SMILES [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Assay Percent Range 0.9932
Quantity 100 g
Formula Weight 291.8
Physical Form Crystalline Powder
Chemical Name or Material Diphenhydramine Hydrochloride
EINECSNumber 205-687-2
Recommended Storage Store at 2°to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.