Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602259 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-259 5 kg Amber Glass Bottle
18-602-253 5 g Amber Glass Bottle
18-602-254 100 g Amber Glass Bottle
18-602-255 500 g Amber Glass Bottle
18-602-258 2.5 kg Amber Glass Bottle
18-602-256 12 kg Amber Glass Bottle
18-602-257 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-602-259 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DI1605KGPL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DI160, 147-24-0, C17H21NO·HCl

Spectrum™ Chemical Diphenhydramine Hydrochloride, USP is an antihistamine that includes sedative attributes to treat allergies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number MFCD00012479
InChI Key PCHPORCSPXIHLZ-UHFFFAOYSA-N
IUPAC Name hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
SMILES [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.