Learn More
Medchemexpress LLC Diphenhydramine (Standard) | 58-73-1 | 98.7% | 50 MG
SDP
Catalog No. 5000406897
Encompass_Preferred
Diphenhydramine (Standard) is the analytical standard of Diphenhydramine, intended for research and analytical applications. It is a first-generation histamine H1-receptor antagonist with anti-cholinergic effects. Diphenhydramine hydrochloride can cross the ovine blood-brain barrier (BBB). This compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • First-generation histamine H1-receptor antagonist.
  • Possesses anti-cholinergic effect.
  • Diphenhydramine hydrochloride can cross the ovine blood-brain barrier (BBB).
  • Commonly used in HPLC, GC, and MS for qualitative, quantitative, and methodological research experiments.
Catalog No. 50-004-06897 Supplier Medchemexpress LLC Supplier No. HYB0303R50MG
Diphenhydramine (Standard) is the analytical standard of Diphenhydramine, intended for research and analytical applications. It is a first-generation histamine H1-receptor antagonist with anti-cholinergic effects. Diphenhydramine hydrochloride can cross the ovine blood-brain barrier (BBB). This compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • First-generation histamine H1-receptor antagonist.
  • Possesses anti-cholinergic effect.
  • Diphenhydramine hydrochloride can cross the ovine blood-brain barrier (BBB).
  • Commonly used in HPLC, GC, and MS for qualitative, quantitative, and methodological research experiments.