Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenhydramine (Standard) | 58-73-1 | MFCD00274173 | 98.7% | 100 MG
SDP

Catalog No. 5000406667
Encompass_Preferred

Diphenhydramine (Standard) is an analytical standard intended for research and analytical applications. It is a first-generation histamine H1-receptor antagonist with an anti-cholinergic effect and can cross the ovine blood-brain barrier (BBB). This compound is supplied as a reference standard assay and is commonly used in qualitative, quantitative, and methodological research experiments.

  • First-generation histamine H1-receptor antagonist
  • Anti-cholinergic effect
  • Can cross the ovine blood-brain barrier (BBB)
  • Used in HPLC, GC, and MS experiments
  • Molecular weight: 255.35
  • Formula: C17H21NO
  • Appearance: liquid
  • Color: colorless to light yellow
  • Classified as an alkaloid
  • Initial source: endogenous metabolite
  • For research use only

Catalog No. 50-004-06667 Supplier Medchemexpress LLC Supplier No. HYB0303R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenhydramine (Standard) is an analytical standard intended for research and analytical applications. It is a first-generation histamine H1-receptor antagonist with an anti-cholinergic effect and can cross the ovine blood-brain barrier (BBB). This compound is supplied as a reference standard assay and is commonly used in qualitative, quantitative, and methodological research experiments.

  • First-generation histamine H1-receptor antagonist
  • Anti-cholinergic effect
  • Can cross the ovine blood-brain barrier (BBB)
  • Used in HPLC, GC, and MS experiments
  • Molecular weight: 255.35
  • Formula: C17H21NO
  • Appearance: liquid
  • Color: colorless to light yellow
  • Classified as an alkaloid
  • Initial source: endogenous metabolite
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.