Learn More
Apexbio Technology LLC Diphenidol HCl 3254-89-5 200mg
SDP
Catalog No. 5000090844
Encompass_Preferred
Diphenidol hydrochloride (CAS 3254-89-5) is a small molecule that acts as a nonselective antagonist of muscarinic acetylcholine receptors targeting subtypes M1 through M4 It also inhibits multiple voltage-gated ion channels including sodium (Na ) potassium (K ) and calcium (Ca ) channels thereby modulating neuronal excitability Diphenidol hydrochloride has demonstrated antiarrhythmic activity and is frequently employed in studies investigating the neurophysiological mechanisms underlying vertigo and the pathways involved in antiemetic responses Its polypharmacological profile makes it a suitable tool for research into neural signaling and emesis modulation
Catalog No. 50-000-90844 Supplier Apexbio Technology LLC Supplier No. B4783200
Diphenidol hydrochloride (CAS 3254-89-5) is a small molecule that acts as a nonselective antagonist of muscarinic acetylcholine receptors targeting subtypes M1 through M4 It also inhibits multiple voltage-gated ion channels including sodium (Na ) potassium (K ) and calcium (Ca ) channels thereby modulating neuronal excitability Diphenidol hydrochloride has demonstrated antiarrhythmic activity and is frequently employed in studies investigating the neurophysiological mechanisms underlying vertigo and the pathways involved in antiemetic responses Its polypharmacological profile makes it a suitable tool for research into neural signaling and emesis modulation