Learn More
Medchemexpress LLC Diphenidol hydrochloride | 3254-89-5 | 99.1% | C21H28ClNO | 1 ML
SDP
Catalog No. 5000432114
Encompass_Preferred
Diphenidol hydrochloride is a non-selective antagonist of muscarinic M1-M4 receptors with anti-arrhythmic activity. It also acts as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells, making it suitable for research in antivertigo and antinausea studies.
  • Non-selective muscarinic M1-M4 receptor antagonist
  • Potent non-specific blocker of voltage-gated Na+, K+, and Ca2+ channels
  • Used in antivertigo and antinausea research
  • High purity at 99.06%
  • Solid, white to off-white appearance
Catalog No. 50-004-32114 Supplier Medchemexpress LLC Supplier No. HYA008210MM/1ML
Diphenidol hydrochloride is a non-selective antagonist of muscarinic M1-M4 receptors with anti-arrhythmic activity. It also acts as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells, making it suitable for research in antivertigo and antinausea studies.
  • Non-selective muscarinic M1-M4 receptor antagonist
  • Potent non-specific blocker of voltage-gated Na+, K+, and Ca2+ channels
  • Used in antivertigo and antinausea research
  • High purity at 99.06%
  • Solid, white to off-white appearance