Learn More
Medchemexpress LLC Diphenidol hydrochloride | 3254-89-5 | 99.1% | C21H28ClNO | 500 MG
SDP
Catalog No. 5000432775
Encompass_Preferred
Diphenidol hydrochloride (Difenidol hydrochloride) is a non-selective muscarinic M1-M4 receptor antagonist with anti-arrhythmic activity. It also acts as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells. This compound can be utilized in the study of antivertigo and antinausea.
  • Non-selective muscarinic M1-M4 receptor antagonist
  • Possesses anti-arrhythmic activity
  • Potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells
  • Useful in the study of antivertigo and antinausea
  • Inhibits sodium currents and produces spinal anesthesia in N2A cells
  • Reduces neuropathic pain and TNF-α overexpression in rats
Catalog No. 50-004-32775 Supplier Medchemexpress LLC Supplier No. HYA0082500MG
Diphenidol hydrochloride (Difenidol hydrochloride) is a non-selective muscarinic M1-M4 receptor antagonist with anti-arrhythmic activity. It also acts as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells. This compound can be utilized in the study of antivertigo and antinausea.
  • Non-selective muscarinic M1-M4 receptor antagonist
  • Possesses anti-arrhythmic activity
  • Potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells
  • Useful in the study of antivertigo and antinausea
  • Inhibits sodium currents and produces spinal anesthesia in N2A cells
  • Reduces neuropathic pain and TNF-α overexpression in rats