Learn More
Medchemexpress LLC Diphenidol hydrochloride | 3254-89-5 | MFCD00151479 | 99.1% | C21H28ClNO | 1 G
SDP
Catalog No. 5000433554
Encompass_Preferred
Diphenidol hydrochloride is a non-selective muscarinic M1-M4 receptor antagonist with anti-arrhythmic activity. It is also a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells. It can be used in the study of antivertigo and antinausea.
  • Acts as a non-selective muscarinic M1-M4 receptor antagonist
  • Exhibits anti-arrhythmic activity
  • Functions as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells
  • Inhibits sodium currents and can produce spinal anesthesia
  • Shown to reduce neuropathic pain and TNF-α overexpression in rat models
  • Demonstrates an inhibitory effect on exercise and morphine-induced vomiting in pica rats
Catalog No. 50-004-33554 Supplier Medchemexpress LLC Supplier No. HYA00821G
Diphenidol hydrochloride is a non-selective muscarinic M1-M4 receptor antagonist with anti-arrhythmic activity. It is also a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells. It can be used in the study of antivertigo and antinausea.
  • Acts as a non-selective muscarinic M1-M4 receptor antagonist
  • Exhibits anti-arrhythmic activity
  • Functions as a potent non-specific blocker of voltage-gated ion channels (Na+, K+, and Ca2+) in neuronal cells
  • Inhibits sodium currents and can produce spinal anesthesia
  • Shown to reduce neuropathic pain and TNF-α overexpression in rat models
  • Demonstrates an inhibitory effect on exercise and morphine-induced vomiting in pica rats