Learn More
Medchemexpress LLC Diphenyl-1-pyrenylphosphine | 110231-30-6 | MFCD00278755 | 95.0% | 386.42 g/mol | C28H19P | 25 MG
SDP
Catalog No. 5000227302
Encompass_Preferred
Diphenyl-1-pyrenylphosphine (DPPP) is a pyrene-based, peroxide-reactive fluorescent probe used to detect hydroperoxides and reactive oxygen species in biochemical assays. Supplied as a solid, it provides sensitive fluorescence readout and is suitable for preparing stock solutions for analytical and cellular studies. Handle protected from light and store at low temperature to preserve stability.
  • Fluorescent peroxide-reactive probe for hydroperoxide detection.
  • Pyrene-based fluorophore with emission around 380 nm for sensitive readout.
  • Supplied as a solid for convenient stock solution preparation.
  • High purity (about 95%) appropriate for analytical applications.
  • Small packaging ideal for low-volume assays (25 MG).
  • Store protected from light and refrigerated to maintain stability.
Catalog No. 50-002-27302 Supplier Medchemexpress LLC Supplier No. HY11815925MG
Diphenyl-1-pyrenylphosphine (DPPP) is a pyrene-based, peroxide-reactive fluorescent probe used to detect hydroperoxides and reactive oxygen species in biochemical assays. Supplied as a solid, it provides sensitive fluorescence readout and is suitable for preparing stock solutions for analytical and cellular studies. Handle protected from light and store at low temperature to preserve stability.
  • Fluorescent peroxide-reactive probe for hydroperoxide detection.
  • Pyrene-based fluorophore with emission around 380 nm for sensitive readout.
  • Supplied as a solid for convenient stock solution preparation.
  • High purity (about 95%) appropriate for analytical applications.
  • Small packaging ideal for low-volume assays (25 MG).
  • Store protected from light and refrigerated to maintain stability.