Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl-2-pyridylmethane 99+%, Thermo Scientific™

Catalog No. AC117430250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117430250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC117430250 Supplier Thermo Scientific Chemicals Supplier No. 117430250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3678-70-4
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
InChI Key MMXYNKLYVRNTCK-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylmethane, pyridine, 2-diphenylmethyl, 2-diphenylmethyl pyridine, 2-benzhydryl-pyridine, pyridine,2-diphenylmethyl, diphenyl-2-pyridylmethane, 98, pyridine, 2-diphenylmethyl-8ci 9ci
PubChem CID 77250
IUPAC Name 2-benzhydrylpyridine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C3=CC=CC=N3

Specifications

Melting Point 58°C to 61°C
Color Orange-Yellow
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 20, II, 321
Formula Weight 245.32
Percent Purity ≥99.0%
Physical Form Crystalline Powder and Crystals
Chemical Name or Material Diphenyl-2-pyridylmethane, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.