Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl 4-Bromo-alpha-chlorobenzylphosphonate 95.0+%, TCI America™
SDP

Catalog No. D38755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38755G 5 g
D387525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38755G Supplier TCI America Supplier No. D38755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 189099-56-7
Molecular Formula C19H15BrClO3P
Molecular Weight (g/mol) 437.654
InChI Key ZSYMFYMYJVSPDH-UHFFFAOYSA-N
Synonym 4-Bromo-alpha-chlorobenzylphosphonic Acid Diphenyl Ester
PubChem CID 44630008
IUPAC Name 1-bromo-4-[chloro(diphenoxyphosphoryl)methyl]benzene
SMILES C1=CC=C(C=C1)OP(=O)(C(C2=CC=C(C=C2)Br)Cl)OC3=CC=CC=C3

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 437.65
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl 4-Bromo-alpha-chlorobenzylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.