Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Diphenyl-4-piperidinomethanol hydrochloride, 98%, Thermo Scientific™

Catalog No. AC124981000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124981000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC124981000 Supplier Thermo Scientific Chemicals Supplier No. 124981000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1798-50-1
MDL Number MFCD00039039
InChI Key LBRAWKGGVUCSOX-UHFFFAOYSA-N
Synonym diphenyl piperidin-4-yl methanol hydrochloride, azacyclonol hydrochloride, azacyclonol hcl, frenquel hydrochloride, azacyclonol, hydrochloride, unii-4bxx0xnp9r, gamma-pipradol hydrochloride, azacyclonol hydrochloride nf, 4bxx0xnp9r, alpha-4-piperidyl benzhydrol hydrochloride
PubChem CID 15722
IUPAC Name diphenyl(piperidin-4-yl)methanol;hydrochloride
SMILES C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 270.0°C to 281.0°C
Color White
Loss on Drying 10% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C18H21NO·HCl
Quantity 100 g
Merck Index 13, 895
Molecular Weight (g/mol) 303.82
Formula Weight 303.82
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α,α-Diphenyl-4-piperidinomethanol hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-284-9
RTECSNumber TN0925000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.