Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Acetaldehyde,MP Biomedicals

Catalog No. ICN222933 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222933 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222933 Supplier MP Biomedicals, Inc Supplier No. 0522293305

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 947-91-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00006972
InChI Key HLLGFGBLKOIZOM-UHFFFAOYSA-N
Synonym diphenylacetaldehyde, diphenylketen, acetaldehyde, diphenyl, alpha-phenylbenzeneacetaldehyde, diphenylethanal, diphenyl-acetaldehyde, benzeneacetaldehyde, .alpha.-phenyl, diphenyl-acetaldehyd, benzeneacetaldehyde, ?-phenyl, diphenylacetoaldehyde
PubChem CID 13696
IUPAC Name 2,2-diphenylacetaldehyde
SMILES O=CC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 196.25
Chemical Name or Material Diphenyl Acetaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.