Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl alpha-Chlorobenzylphosphonate 98.0+%, TCI America™
SDP

Catalog No. D38245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38245G 5 g
D38241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38245G Supplier TCI America Supplier No. D38245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58263-67-5
Molecular Formula C19H16ClO3P
Molecular Weight (g/mol) 358.76
MDL Number MFCD00159438
InChI Key LTRZPFCYQMGZGD-UHFFFAOYNA-N
Synonym alpha-Chlorobenzylphosphonic Acid Diphenyl Ester
PubChem CID 277711
IUPAC Name diphenyl [chloro(phenyl)methyl]phosphonate
SMILES ClC(C1=CC=CC=C1)P(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color White
Boiling Point 210°C
Quantity 5 g
Formula Weight 358.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl alpha-Chlorobenzylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.