Learn More
Medchemexpress LLC HY-D0970 5g Medchemexpress, Diphenyl Blue CAS:72-57-1 Purity:>98%
SDP
Catalog No. 501933217
Encompass_Preferred
Medchemexpress, HY-D0970 5g Diphenyl Blue CAS:72-57-1 Diphenyl Blue (Direct Blue 14) is used as reference dye for group of azo dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3217 Supplier Medchemexpress LLC Supplier No. HYD09705G
Medchemexpress, HY-D0970 5g Diphenyl Blue CAS:72-57-1 Diphenyl Blue (Direct Blue 14) is used as reference dye for group of azo dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.