Learn More
Medchemexpress LLC Diphenyl Blue | 72-57-1 | 39.0% | 25 G
SDP
Catalog No. 5000414294
Encompass_Preferred
Diphenyl Blue (Trypan Blue) is a cell-active dye, most commonly used for the identification of dead cells. It is frequently employed to assess cell membrane integrity and cell viability. This dye stains deactivated cells or those with incomplete cell membranes blue, while normal living cells with intact membranes do not absorb it. Macrophages, however, can phagocytose Diphenyl Blue, allowing its use as a living stain for these cells.
  • Cell-active dye
  • Most commonly used dye for the identification of dead cells
  • Used to test cell membrane integrity and cell viability
  • Can be used as a living stain for macrophages
Catalog No. 50-004-14294 Supplier Medchemexpress LLC Supplier No. 25GHYD0970
Diphenyl Blue (Trypan Blue) is a cell-active dye, most commonly used for the identification of dead cells. It is frequently employed to assess cell membrane integrity and cell viability. This dye stains deactivated cells or those with incomplete cell membranes blue, while normal living cells with intact membranes do not absorb it. Macrophages, however, can phagocytose Diphenyl Blue, allowing its use as a living stain for these cells.
  • Cell-active dye
  • Most commonly used dye for the identification of dead cells
  • Used to test cell membrane integrity and cell viability
  • Can be used as a living stain for macrophages