Learn More
Apexbio Technology LLC DIPHENYL-DISULFIDE-1G
SDP
Catalog No. 5000568223
Encompass_Preferred
Diphenyl disulfide (CAS No 882-33-7) is widely utilized in biomedical research particularly in the study of oxidative stress and sulfur-containing compound mechanisms Originating as an organosulfur compound it functions by modulating redox signaling pathways and altering thiol-disulfide exchange reactions In vitro its activity is observed in nanomolar to low micromolar ranges making it effective in various biochemical assays Commonly it is used in cellular models to investigate oxidative damage responses and in animal studies to evaluate potential therapeutic effects against oxidative stress-related conditions It also serves as a tool in drug screening processes to identify agents affecting disulfide bond dynamics Experimental concentrations typically depend on the specific research design ensuring adaptability across different experimental setups
Catalog No. 50-005-68223 Supplier Apexbio Technology LLC Supplier No. C68081000
Diphenyl disulfide (CAS No 882-33-7) is widely utilized in biomedical research particularly in the study of oxidative stress and sulfur-containing compound mechanisms Originating as an organosulfur compound it functions by modulating redox signaling pathways and altering thiol-disulfide exchange reactions In vitro its activity is observed in nanomolar to low micromolar ranges making it effective in various biochemical assays Commonly it is used in cellular models to investigate oxidative damage responses and in animal studies to evaluate potential therapeutic effects against oxidative stress-related conditions It also serves as a tool in drug screening processes to identify agents affecting disulfide bond dynamics Experimental concentrations typically depend on the specific research design ensuring adaptability across different experimental setups