Learn More
Medchemexpress LLC Diphenyl disulfide | 882-33-7 | MFCD00003065 | 100.0% | 218.35 g/mol | C12H10S2 | 5g
SDP
Catalog No. 5000266802
Encompass_Preferred
Diphenyl disulfide is an endogenous metabolite Diphenyl disulfide is a pharmaceutical intermediate Diphenyl disulfide can be coupled with aryl iodide to synthesize asymmetric aryl sulfide[1][2]
Catalog No. 50-002-66802 Supplier Medchemexpress LLC Supplier No. HYY11775G
Diphenyl disulfide is an endogenous metabolite Diphenyl disulfide is a pharmaceutical intermediate Diphenyl disulfide can be coupled with aryl iodide to synthesize asymmetric aryl sulfide[1][2]