Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Iodonium-2-carboxylate, MP Biomedicals

Catalog No. ICN227676 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227676 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227676 Supplier MP Biomedicals, Inc Supplier No. 0522767625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109545-72-4
Molecular Formula C13H9IO2
Molecular Weight (g/mol) 324.117
InChI Key LUGVQQXOGHCZNN-UHFFFAOYSA-N
Synonym diphenyliodonium-2-carboxylate, iodonium, 2-carboxyphenyl phenyl-, inner salt, 2-carboxylatophenyl phenyliodonium, 2-carboxyphenyl phenyliodonium hydroxide inner salt, iodonium, 2-carboxyphenyl phenyl-,hydroxide,innersalt, diphenyliodonium-2-carboxylate monohydrate, 98+%, 2-carboxylatophenyl phenyl iodanium, iodonium, hydroxide, inner salt, iodonium, 2-carboxyphenyl phenyl-, hydroxide, inner salt, iodonium, o-carboxyphenyl phenyl-, hydroxide, inner salt
PubChem CID 73883
IUPAC Name 2-phenyliodoniobenzoate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2C(=O)[O-]

Specifications

Quantity 10 g
Formula Weight 342.13
Chemical Name or Material Diphenyl Iodonium-2-Carboxylate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.