Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate 95.0+%, TCI America™
SDP

Catalog No. D37095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37095G 5 g
D37091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37095G Supplier TCI America Supplier No. D37095G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 367508-01-8
Molecular Formula C16H18NO5P
Molecular Weight (g/mol) 335.30
MDL Number MFCD11042674
InChI Key CFSFGLDTIDFDMT-UHFFFAOYSA-N
Synonym (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diphenyl Ester
PubChem CID 12030864
IUPAC Name diphenyl {[methoxy(methyl)carbamoyl]methyl}phosphonate
SMILES CON(C)C(=O)CP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 335.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.