Learn More
Medchemexpress LLC Diphenyl Phosphate | 838-85-7 | 250.19 | 1 ML
SDP
Catalog No. 5000445770
Encompass_Preferred
Diphenyl Phosphate is a chemical compound that inhibits growth and energy metabolism in zebrafish and mice in a sex-specific manner. It achieves this by inhibiting the activity of succinate dehydrogenase (SDH), reducing the expression of carnitine palmitoyltransferase 1 (CPT1), and disrupting the integrity of the mitochondrial membrane. This product is intended for research use only, specifically in studies related to metabolic diseases.
  • Inhibits growth and energy metabolism
  • Inhibits succinate dehydrogenase (SDH) and reduces CPT1 expression
  • Disrupts mitochondrial membrane integrity
  • Useful in research on metabolic diseases
  • Causes disturbances in lipid metabolism in male mice
  • Retards growth in male zebrafish by inhibiting energy output
Catalog No. 50-004-45770 Supplier Medchemexpress LLC Supplier No. HYW00815110MM/1ML
Diphenyl Phosphate is a chemical compound that inhibits growth and energy metabolism in zebrafish and mice in a sex-specific manner. It achieves this by inhibiting the activity of succinate dehydrogenase (SDH), reducing the expression of carnitine palmitoyltransferase 1 (CPT1), and disrupting the integrity of the mitochondrial membrane. This product is intended for research use only, specifically in studies related to metabolic diseases.
  • Inhibits growth and energy metabolism
  • Inhibits succinate dehydrogenase (SDH) and reduces CPT1 expression
  • Disrupts mitochondrial membrane integrity
  • Useful in research on metabolic diseases
  • Causes disturbances in lipid metabolism in male mice
  • Retards growth in male zebrafish by inhibiting energy output