Learn More
Medchemexpress LLC Diphenyl Phosphate-d10 | 1477494-97-5 | 260.25 | 1 MG
SDP
Catalog No. 5000445691
Encompass_Preferred
Diphenyl Phosphate-d10 (DPhP-d10) is a deuterium-labeled Diphenyl Phosphate that can inhibit growth and energy metabolism of zebrafish in a sex-specific manner. This compound is suitable as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, and LC-MS. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-45691 Supplier Medchemexpress LLC Supplier No. HYW008151S1MG
Add to Cart
Add to Cart
Diphenyl Phosphate-d10 (DPhP-d10) is a deuterium-labeled Diphenyl Phosphate that can inhibit growth and energy metabolism of zebrafish in a sex-specific manner. This compound is suitable as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, and LC-MS. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS