Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diphenyl Phosphoryl Azide

Catalog No. 1787565KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-875-65KG 5 kg
17-875-625G 25 g
17-875-6100G 100 g
17-875-65G 5 g
17-875-6500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 17-875-65KG Supplier Sigma Aldrich Supplier No. 1787565KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as the activating agent in the preparation of macrocyclic lactams and of an aldose reductase inhibitor.

Specifications

CAS 26386-88-9
Density 1.277 g/mL at 25°C
Boiling Point 157°C (0.17 mmHg, lit.)
Molecular Formula C12H10N3O3P
Refractive Index n20/D 1.551
Linear Formula (C6H5O)2P(O)N3
Quantity 5 kg
MDL Number MFCD00001987
Synonym DPPA; Phosphoric acid diphenyl ester azide
Molecular Weight (g/mol) 275.2 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.