Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diphenyl phosphoryl chloride

Catalog No. 344249500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-424-9500G 500 g
34-424-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-424-9500G Supplier Sigma Aldrich Supplier No. 344249500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Diphenyl phosphoryl chloride is a diaryl phosphoryl derivative. it is formed during the preparation of monophenyl phosphoryl chloride via reaction of phenol and phosphorus oxychloride.diphenyl phosphoryl chloride may be used:
  • In the preparation of guanosine-adenosine-5α,5α-triphosphate, guanosine-cytidine-5α,5α-triphosphate, guanosine-uridine-5α,5α-triphosphate, diguanosine-5α,5α-diphosphate and diguanosine-5α,5α-triphosphate
  • As phosphorylating agent for the synthesis of 2-deoxy-d-galactose-3 and -6 phosphoric acids
  • In the synthesis of diaryl phosphoryl derivatives of alkylene dithiophosphates

Specifications

CAS 2524-64-3
Density 1.296 g/mL (at 25°C (literature))
Boiling Point 314°C to 316°C (272 mmHg)
Molecular Formula C12H10ClO3P
Refractive Index n20/D 1.55 (literature)
Linear Formula (C6H5O)2 POCl
MDL Number MFCD00003030
Quantity 500 g
Synonym Diphenyl chlorophosphate; Diphenyl phosphorochloridate
Molecular Weight (g/mol) 268.63
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.