Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenyl phthalate-3,4,5,6-d4 | 1398065-61-6 | MFCD22565776 | >98.0% | 10 MG
SDP

Catalog No. 5000298854
Encompass_Preferred

Diphenyl phthalate-3,4,5,6-d4 is a deuterium-labeled analytical standard of diphenyl phthalate supplied for research use. It is used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is provided as a white to off-white solid with recommended low-temperature storage.

  • Deuterium labeled at positions 3,4,5,6 on the phthalate ring.
  • Molecular weight 322.35 and formula C20H10D4O4.
  • CAS number 1398065-61-6 for unambiguous identification.
  • Suitable as an internal standard or tracer for NMR, GC-MS, and LC-MS analyses.
  • Available in small analytical quantities (1 mg-100 mg) with recommended cold storage.

Catalog No. 50-002-98854 Supplier Medchemexpress LLC Supplier No. HYB1966S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyl phthalate-3,4,5,6-d4 is a deuterium-labeled analytical standard of diphenyl phthalate supplied for research use. It is used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is provided as a white to off-white solid with recommended low-temperature storage.

  • Deuterium labeled at positions 3,4,5,6 on the phthalate ring.
  • Molecular weight 322.35 and formula C20H10D4O4.
  • CAS number 1398065-61-6 for unambiguous identification.
  • Suitable as an internal standard or tracer for NMR, GC-MS, and LC-MS analyses.
  • Available in small analytical quantities (1 mg-100 mg) with recommended cold storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.